Learn More
Novus Biologicals™ TRPA1 Peptide

Specifications
Specifications
| Gene ID (Entrez) | 8989 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-40763 |
| Gene Alias | ANKTM1ankyrin-like with transmembrane domains 1, Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, transient receptor potential cation channel subfamily A member 1, transient receptor potential cation channel, subfamily A, member 1 |
| Gene Symbol | TRPA1 |
| Quantity | 0.1 mg |
| Research Category | Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems |
| Storage Buffer | Peptide dissolved in dH2O. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.