Learn More
Aviva Systems Biology Corp TRPC3 Antibody - N-terminal region (ARP58715_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58715_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRPC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human TRPC3. Alternate names: TRP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62399