Learn More
Aviva Systems Biology Corp TRPC4AP Antibody - middle region (ARP55313_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55313_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRPC4AP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRPC4AP. Alternate names: C20orf188, TRRP4AP, TRUSSApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61464