Learn More
Medchemexpress LLC Trpc5-in-1 | 2265215-18-5 | 99.0% | C20H16N4O | 250 MG
SDP
Catalog No. 5000352831
Encompass_Preferred
TRPC5-IN-1 (Compound 6j) is a selective TRPC5 inhibitor. This solid, white to off-white compound demonstrates 50.5% inhibition for TRPC5 at 3 μM and is suitable for research related to chronic kidney disease (CKD). It is intended for research use only.
  • Selective TRPC5 inhibitor
  • Suitable for chronic kidney disease (CKD) research
  • Solid, white to off-white appearance
  • 50.5% inhibition for TRPC5 at 3 μM
Catalog No. 50-003-52831 Supplier Medchemexpress LLC Supplier No. 250MGHY145150
TRPC5-IN-1 (Compound 6j) is a selective TRPC5 inhibitor. This solid, white to off-white compound demonstrates 50.5% inhibition for TRPC5 at 3 μM and is suitable for research related to chronic kidney disease (CKD). It is intended for research use only.
  • Selective TRPC5 inhibitor
  • Suitable for chronic kidney disease (CKD) research
  • Solid, white to off-white appearance
  • 50.5% inhibition for TRPC5 at 3 μM