Learn More
Aviva Systems Biology Corp TRPM2 Antibody - N-terminal region (ARP44380_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRPM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRPM2. Alternate names: EREG1, KNP3, LTRPC2, MGC133383, NUDT9H, NUDT9L1, TRPC7ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53418