Learn More
Aviva Systems Biology Corp TRPM3 Antibody - N-terminal region (ARP35609_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35609_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRPM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRPM3. Alternate names: GON-2, LTRPC3, MLSN2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53014