Learn More
Aviva Systems Biology Corp TRPM4 Antibody - N-terminal region (ARP35268_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35268_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRPM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRPM4. Alternate names: FLJ20041, TRPM4B, PFHB1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-52998