Learn More
ABclonal Technology TRPM4 Rabbit mAb
SDP
Catalog No. 5000012778
Encompass_Preferred
The protein encoded by this gene is a calcium-activated nonselective ion channel that mediates transport of monovalent cations across membranes thereby depolarizing the membrane The activity of the encoded protein increases with increasing intracellular calcium concentration but this channel does not transport calcium
Catalog No. 50-000-12778 Supplier ABclonal Technology Supplier No. A2551120UL
The protein encoded by this gene is a calcium-activated nonselective ion channel that mediates transport of monovalent cations across membranes thereby depolarizing the membrane The activity of the encoded protein increases with increasing intracellular calcium concentration but this channel does not transport calcium