Learn More
Medchemexpress LLC HY-112430 10mg Medchemexpress, TRPM8 antagonist 2 CAS:259674-19-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11243010MG
Encompass_Preferred
Medchemexpress, HY-112430 10mg TRPM8 antagonist 2 CAS:259674-19-6 TRPM8 antagonist 2 is a potent and selective TRPM8 antagonist, with an IC50 of 0.2 nM, used in the research of neuropathic pain syndromes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9090