Learn More
Izon Science Usa Ltd TRPS REAGENT KIT
SDP
Catalog No. NC2389299
Encompass
Improve the performance, consistency and ease of use of your TRPS instrument with the new TRPS Reagent Kit. Now shipping with an extended shelf life.Izon's Reagent Kit makes TRPS analysis faster and more repeatable. The key ingredient is Izon Coating Solution (ICS), which minimises non-specific binding of particles to the nanopore surface (particularly in combination with qEV purification).Reliable and consistent results; measurements on different nanopores are comparable and accurateStable operation, reduces non-specific binding to nanoporeContaminant-free solutionsExtended shelf lifeHigher throughputThe Reagent Kit Contains:1x - Wetting Solution Concentrate1x - 8.5g Coating Solution (ICS)4x - PBS Tablets 20x - 13mm x 0.22μm Syringe Filter4x - 25mm x 0.45μm Syringe Filter1x - Reagent Kit Technical NoteProduct code: RK3
Catalog No. NC2389299 Supplier Izon Science Usa Ltd Supplier No. RK3
Improve the performance, consistency and ease of use of your TRPS instrument with the new TRPS Reagent Kit. Now shipping with an extended shelf life.Izon's Reagent Kit makes TRPS analysis faster and more repeatable. The key ingredient is Izon Coating Solution (ICS), which minimises non-specific binding of particles to the nanopore surface (particularly in combination with qEV purification).Reliable and consistent results; measurements on different nanopores are comparable and accurateStable operation, reduces non-specific binding to nanoporeContaminant-free solutionsExtended shelf lifeHigher throughputThe Reagent Kit Contains:1x - Wetting Solution Concentrate1x - 8.5g Coating Solution (ICS)4x - PBS Tablets 20x - 13mm x 0.22μm Syringe Filter4x - 25mm x 0.45μm Syringe Filter1x - Reagent Kit Technical NoteProduct code: RK3