Learn More
Alkali Scientific Trptic Soy Agar w/5% Sheeps Blood and Esculin 20mL, 10/Pk
SDP
Catalog No. 5000663269
Encompass_Preferred
Tryptic Soy Agar with 5% Sheep’s Blood and Esculin, 20mL Plate, 10/Pk is an enriched differential culture medium designed for the cultivation and preliminary identification of fastidious microorganisms. The addition of 5% sheep’s blood supports the growth of demanding bacterial species while allowing observation of hemolytic activity around colonies. Esculin in the formulation enables differentiation of organisms capable of hydrolyzing esculin, producing a dark reaction in the surrounding medium. This combination is particularly useful in detecting and differentiating Enterococci and Group D Streptococci. Each plate contains 20mL of sterile agar prepared for immediate use in microbiology laboratories. The medium supports diagnostic, environmental, and quality control testing workflows requiring reliable differentiation and organism recovery.
Catalog No. 50-006-63269 Supplier Alkali Scientific Supplier No. TSAP05
Tryptic Soy Agar with 5% Sheep’s Blood and Esculin, 20mL Plate, 10/Pk is an enriched differential culture medium designed for the cultivation and preliminary identification of fastidious microorganisms. The addition of 5% sheep’s blood supports the growth of demanding bacterial species while allowing observation of hemolytic activity around colonies. Esculin in the formulation enables differentiation of organisms capable of hydrolyzing esculin, producing a dark reaction in the surrounding medium. This combination is particularly useful in detecting and differentiating Enterococci and Group D Streptococci. Each plate contains 20mL of sterile agar prepared for immediate use in microbiology laboratories. The medium supports diagnostic, environmental, and quality control testing workflows requiring reliable differentiation and organism recovery.