Learn More
Cayman Chemical TRPV1-Tat (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502914963
Encompass_Preferred
A peptide TRPV1 antagonist; inhibits heat- or PMA-induced currents in whole-cell patch-clamp assays using primary mouse dorsal root ganglia at 200 µM; increases the hind paw mechanical pain threshold in rats at 10 or 30 µM
Catalog No. 50-291-4963 Supplier Cayman Chemical Supplier No. 3695510 MG
A peptide TRPV1 antagonist; inhibits heat- or PMA-induced currents in whole-cell patch-clamp assays using primary mouse dorsal root ganglia at 200 µM; increases the hind paw mechanical pain threshold in rats at 10 or 30 µM