Learn More
Medchemexpress LLC TRPV4 agonist-1 free 10mg | 2314467-59-7 | 10 MG
SDP
Catalog No. 5000229475
Encompass_Preferred
TRPV4 agonist-1 free base is a small-molecule agonist of the transient receptor potential vanilloid 4 (TRPV4) ion channel. It activates human TRPV4 with an EC50 of 60 nM in calcium mobilization assays and is supplied for research use only.
  • Activates human TRPV4 with EC50 of 60 nM in calcium mobilization assay.
  • Measured by intracellular Ca2+ increase in HEK293 cells using Fura-2 AM probe.
  • Molecular formula C25H22F2N4O2; molecular weight 448.46 g/mol.
  • High reported purity (~99.7%).
  • Available in multiple mg-scale sizes for laboratory research applications.
  • Intended for laboratory research use only, not for human use.
Catalog No. 50-002-29475 Supplier Medchemexpress LLC Supplier No. HY11440010MG
TRPV4 agonist-1 free base is a small-molecule agonist of the transient receptor potential vanilloid 4 (TRPV4) ion channel. It activates human TRPV4 with an EC50 of 60 nM in calcium mobilization assays and is supplied for research use only.
  • Activates human TRPV4 with EC50 of 60 nM in calcium mobilization assay.
  • Measured by intracellular Ca2+ increase in HEK293 cells using Fura-2 AM probe.
  • Molecular formula C25H22F2N4O2; molecular weight 448.46 g/mol.
  • High reported purity (~99.7%).
  • Available in multiple mg-scale sizes for laboratory research applications.
  • Intended for laboratory research use only, not for human use.