Learn More
Medchemexpress LLC TRPV4 agonist-1 free | 50MG
SDP
Catalog No. 5000307601
Encompass_Preferred
TRPV4 agonist-1 free base is a transient receptor potential vanilloid 4 (TRPV4) agonist with an EC50 of 60 nM in the hTRPV4 Ca2 assay[1]
Catalog No. 50-003-07601 Supplier Medchemexpress LLC Supplier No. HY11440050MG
TRPV4 agonist-1 free base is a transient receptor potential vanilloid 4 (TRPV4) agonist with an EC50 of 60 nM in the hTRPV4 Ca2 assay[1]