Learn More
Medchemexpress LLC HY-114400 5mg Medchemexpress, TRPV4 agonist-1 (free base) CAS:2314467-59-7 Purity:>98%
SDP
Catalog No. 502080871
Encompass_Preferred
Medchemexpress, HY-114400 5mg TRPV4 agonist-1 (free base) CAS:2314467-59-7 TRPV4 agonist-1 free base is a transient receptor potential vanilloid 4 (TRPV4) agonist with an EC50 of 60 nM in the hTRPV4 Ca2+ assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0871 Supplier Medchemexpress LLC Supplier No. HY1144005MG
Medchemexpress, HY-114400 5mg TRPV4 agonist-1 (free base) CAS:2314467-59-7 TRPV4 agonist-1 free base is a transient receptor potential vanilloid 4 (TRPV4) agonist with an EC50 of 60 nM in the hTRPV4 Ca2+ assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.