Learn More
Aviva Systems Biology Corp TRPV4 Antibody - middle region (ARP35330_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35330_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRPV4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRPV4. Alternate names: OTRPC4, TRP12, VR-OAC, VRL-2, VRL2, VROAC, CMT2C, SPSMA, HMSN2C, SSQTL1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55699