Learn More
Aviva Systems Biology Corp TRPV5 Antibody - N-terminal region (ARP49458_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49458_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRPV5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRPV5. Alternate names: CAT2, ECAC1, OTRPC3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60090