Learn More
Aviva Systems Biology Corp Trpv6 Antibody - middle region (ARP35284_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35284_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Trpv6. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: CaT1, Ecac2, Otrpc3ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-458