Learn More
Worthington Biochemical Corporation TRPYSIN TPCK TREATED
SDP
Catalog No. NC9548088
Encompass
Chromatographically purified trypsin treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity according to Kostka, V., and Carpenter, F.H.: JBC, 239, 1799 (1964).
Catalog No. NC9548088 Supplier Worthington Biochemical Corporation Supplier No. LS003750
Chromatographically purified trypsin treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity according to Kostka, V., and Carpenter, F.H.: JBC, 239, 1799 (1964).