Learn More
Grainger TRS(F) 6.35mm to TRS(F) 6.35mm Coupler
Catalog No. 502752521
Encompass_Preferred
The Grainger 14C222 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and allows use of 1/4-inch drive tools with 3/8-inch drive socket accessories. This socket adapter has a black oxide finish to resist corrosion. It measures 1-1/2 inches long and weighs 0.8 ounces. The 14C222 socket adapter is ideal for mechanics to increase tool compatibility between different drive sizes.
Catalog No. 50-275-2521 Supplier Grainger Supplier No. 14C222
The Grainger 14C222 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and allows use of 1/4-inch drive tools with 3/8-inch drive socket accessories. This socket adapter has a black oxide finish to resist corrosion. It measures 1-1/2 inches long and weighs 0.8 ounces. The 14C222 socket adapter is ideal for mechanics to increase tool compatibility between different drive sizes.