Learn More
Aviva Systems Biology Corp TRSPAP1 Antibody - N-terminal region (ARP40881_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40881_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRSPAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRSPAP1. Alternate names: FLJ20503, PRO1902, RP4-669K10.4, SECP43, TRSPAP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57651