Learn More
Medchemexpress LLC Trt-Gly-OH | 5893-05-0 | MFCD00004276 | 95.2% | 317.39 | C21H19NO2 | 5 G
SDP
Catalog No. 5000283554
Encompass_Preferred
Trt-Gly-OH is a trityl-protected glycine derivative commonly used as an amino acid building block in peptide synthesis and organic chemistry. The trityl (Trt) group provides temporary protection of the amino functionality to minimize side reactions during coupling and deprotection steps. The compound is supplied as a solid appropriate for laboratory-scale research use.
  • Trityl-protected glycine for temporary amino protection.
  • Suitable for peptide synthesis and organic intermediate preparation.
  • Provided as a solid in laboratory pack sizes, for example 5 G.
  • Typical purity around 95.2%, suitable for research applications.
  • Molecular weight approximately 317.39, molecular formula C21H19NO2.
Catalog No. 50-002-83554 Supplier Medchemexpress LLC Supplier No. HYW1295855G
Trt-Gly-OH is a trityl-protected glycine derivative commonly used as an amino acid building block in peptide synthesis and organic chemistry. The trityl (Trt) group provides temporary protection of the amino functionality to minimize side reactions during coupling and deprotection steps. The compound is supplied as a solid appropriate for laboratory-scale research use.
  • Trityl-protected glycine for temporary amino protection.
  • Suitable for peptide synthesis and organic intermediate preparation.
  • Provided as a solid in laboratory pack sizes, for example 5 G.
  • Typical purity around 95.2%, suitable for research applications.
  • Molecular weight approximately 317.39, molecular formula C21H19NO2.