Learn More
Cayman Chemical TRtIapIn LQtrIfluoroACTAt 10mg
SDP
Catalog No. 502917830
Encompass_Preferred
A peptide derivative of tertiapin; inhibits heteromultimeric potassium channels composed of GIRK1 and GIRK4, as well as Kir1.1; suppresses Purkinje cell pause response to a conditional stimulus in ferrets
Catalog No. 50-291-7830 Supplier Cayman Chemical Supplier No. 3749610 MG
A peptide derivative of tertiapin; inhibits heteromultimeric potassium channels composed of GIRK1 and GIRK4, as well as Kir1.1; suppresses Purkinje cell pause response to a conditional stimulus in ferrets