Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Tru-Q 4 (5% Tier) Reference Standard, Unit Size: 1 µg, Concentration: 50 ng/µl
Catalog No. NC2739751
Encompass
The Tru-Q DNA Reference Standards are a highly-characterized, biologically-relevant quality control material used to assess the performance of NGS assays that detect somatic mutations.
Catalog No. NC2739751 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. HD731
The Tru-Q DNA Reference Standards are a highly-characterized, biologically-relevant quality control material used to assess the performance of NGS assays that detect somatic mutations.