Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer AEMC 514 Clamp-On Meter, 0.05 to 1000 A(rms)

Catalog No. 50992612 Shop All Cole-Parmer Products
Encompass

AC and AC/DC Clamp-On Meters
Auto-Ranging Models

  • Models AE211754, AE211721, AE211766 and AE211722 are compact in size and measure to 400A and 600V
  • Models AE211767, AE211768 and AE211770 are standard size clamp-ons and measure to 1000A and 1000V
  • Full functions: Aac, Adc, Vac, Vdc, Ohms, continuity with beeper, frequency, diode test and hold or 1ms peak
  • Several models offer DC amp measurements for non-linear applications
  • 42-segment analog bargraph
  • Large easy-to-read 4000-count display

Catalog No. 50-992-612 Supplier Cole-Parmer Supplier No. 2000269
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC and AC/DC Clamp-On Meters
Auto-Ranging Models

  • Models AE211754, AE211721, AE211766 and AE211722 are compact in size and measure to 400A and 600V
  • Models AE211767, AE211768 and AE211770 are standard size clamp-ons and measure to 1000A and 1000V
  • Full functions: Aac, Adc, Vac, Vdc, Ohms, continuity with beeper, frequency, diode test and hold or 1ms peak
  • Several models offer DC amp measurements for non-linear applications
  • 42-segment analog bargraph
  • Large easy-to-read 4000-count display

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.