Learn More
Chemglass Life Sciences Compact Tru-Stir Shaft Coupling fits 12.7mm Shaft OD for use with Jacobs Chuck
SDP
Catalog No. 501531416
Encompass_Preferred
Compact Tru-Stir Shaft Coupling fits 12.7mm Shaft OD for use with Jacobs Chuck. Similar to CG-2044 but the overall height is 1 shorter. The unique design of Chemglass's Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical Stainless Steel spring permitting up to 1/8 of offset misalignment. Coupling is rated for 21 in-lbs of torque without the backlash normally associated with common coil springs.
Catalog No. 50-153-1416 Supplier Chemglass Life Sciences Supplier No. CG2044C11
Compact Tru-Stir Shaft Coupling fits 12.7mm Shaft OD for use with Jacobs Chuck. Similar to CG-2044 but the overall height is 1 shorter. The unique design of Chemglass's Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical Stainless Steel spring permitting up to 1/8 of offset misalignment. Coupling is rated for 21 in-lbs of torque without the backlash normally associated with common coil springs.