Learn More
Chemglass Life Sciences 12.7mm Tru-Stir Stirrer Shaft Coupling, 3/8" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck
SDP
Catalog No. CG204402
Encompass_Preferred
12.7mm Tru-Stir Stirrer Shaft Coupling, 3/8" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 12.7mm (OD) stirrer shafts. Upper adapters permit the coupling to be used with a standard Jacobs style 3-jaw chuck. Male Upper Coupling is 3/8". Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.
Catalog No. CG204402 Supplier Chemglass Life Sciences Supplier No. CG204402
12.7mm Tru-Stir Stirrer Shaft Coupling, 3/8" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 12.7mm (OD) stirrer shafts. Upper adapters permit the coupling to be used with a standard Jacobs style 3-jaw chuck. Male Upper Coupling is 3/8". Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.