Learn More
Chemglass Life Sciences 10mm Tru-Stir Stirrer Shaft Coupling, 3/8" Upper Female Coupling, For Use with CG-2021 Thru CG-2025
SDP
Catalog No. CG204412
Encompass_Preferred
10mm Tru-Stir Stirrer Shaft Coupling, 3/8" Upper Female Coupling, For Use with CG-2021 Thru CG-2025. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 10mm (OD) stirrer shafts. Female Upper Coupling is 3/8". For use with CG-2021 thru CG-2025 series of motors and stirrers. Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.
Catalog No. CG204412 Supplier Chemglass Life Sciences Supplier No. CG204412
10mm Tru-Stir Stirrer Shaft Coupling, 3/8" Upper Female Coupling, For Use with CG-2021 Thru CG-2025. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 10mm (OD) stirrer shafts. Female Upper Coupling is 3/8". For use with CG-2021 thru CG-2025 series of motors and stirrers. Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.