Learn More
Chemglass Life Sciences 10mm Tru-Stir Stirrer Shaft Coupling, 12mm Upper Female Coupling
SDP
Catalog No. CG204440
Encompass_Preferred
10mm Tru-Stir Stirrer Shaft Coupling, 12mm Upper Female Coupling. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 10mm (OD) stirrer shafts. Female Upper Coupling is 12mm. Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.
Catalog No. CG204440 Supplier Chemglass Life Sciences Supplier No. CG204440
10mm Tru-Stir Stirrer Shaft Coupling, 12mm Upper Female Coupling. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 10mm (OD) stirrer shafts. Female Upper Coupling is 12mm. Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.