Learn More
Chemglass Life Sciences 19mm Tru-Stir Stirrer Shaft Coupling, 3/4" Upper Female Coupling, For Use with CG-2033-B-25, CG-2033-B-38, & CG-2033-B-39
SDP
Catalog No. CG204430
Encompass_Preferred
19mm Tru-Stir Stirrer Shaft Coupling, 3/4" Upper Female Coupling, For Use with CG-2033-11. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 19mm (OD) stirrer shafts. Female Upper Coupling is 3/4". For Use with CG-2033-B overhead stirrers. Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.
Catalog No. CG204430 Supplier Chemglass Life Sciences Supplier No. CG204430
19mm Tru-Stir Stirrer Shaft Coupling, 3/4" Upper Female Coupling, For Use with CG-2033-11. The design of our Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 19mm (OD) stirrer shafts. Female Upper Coupling is 3/4". For Use with CG-2033-B overhead stirrers. Overall length of the coupling is 4¿. Supplied complete with an allen wrench set.