Learn More
Chemglass Life Sciences 10mm Tru-Stir Stirrer Shaft Coupling, Tool-Free, 5/16" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck
SDP
Catalog No. CG204301
Encompass_Preferred
10mm Tru-Stir Stirrer Shaft Coupling, Tool-Free, 5/16" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck. The unique design of our Tool-Free Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical stainless steel spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs.Lower portion of coupling has an anodized aluminum compression nut that secures a 10mm diameter stirrer shaft via a neoprene insert. This unique design allows a Tool-Free connection between the stirrer shaft and coupling. Upper adapter is 5/16¿ diameter, which permits the coupling to be used with a standard Jacobs style 3-jaw chuck. Overall length of the coupling is 4-3/4¿.
Catalog No. CG204301 Supplier Chemglass Life Sciences Supplier No. CG204301
10mm Tru-Stir Stirrer Shaft Coupling, Tool-Free, 5/16" Upper Male Coupling, For Use with 3-Jaw Jacobs Chuck. The unique design of our Tool-Free Tru-Stir stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical stainless steel spring permitting up to 1/4¿ of offset misalignment and 45¿ of angular misalignment. Coupling is rated for 24 in-lbs of torque without the backlash normally associated with common coil springs.Lower portion of coupling has an anodized aluminum compression nut that secures a 10mm diameter stirrer shaft via a neoprene insert. This unique design allows a Tool-Free connection between the stirrer shaft and coupling. Upper adapter is 5/16¿ diameter, which permits the coupling to be used with a standard Jacobs style 3-jaw chuck. Overall length of the coupling is 4-3/4¿.