Learn More
Aviva Systems Biology Corp TRUB1 Antibody - C-terminal region (ARP62931_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62931_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRUB1. It was validated on Western Blot. . Alternate names: PUS4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-287-811