Learn More
Electron Microscopy Sciences TruBond™ 360 Adhesion Slides, Blue
SDP
Supplier:  Electron Microscopy Sciences 63701B1
Encompass_Preferred
Adhesive slides have proven performance under high temperature and pH condition. These adhesive slides, including the hydrophilic TruBond 360, work well with automatic IHC applications. EMS is confident that the quality of these slides will exceed your expectations. For problems with tissue loss try our TruBond 380.
Color: Blue
Catalog No. 50-340-42