Learn More
Newcomer Supply Trubond 380 Adhesive Microscope Slides, Blue (1,000 slides/case)
SDP
Catalog No. NC2101954
Encompass
The surface of the TruBond 380 microscope slide is hydrophilic allowing the tissued to be refloated in the water bath for repositioning the tissue section and allowing the microscope slide to be used on all automated IHC platforms. They are especially useful in preventing tissue loss when performing high temperature antigen retrieval or enzyme digestion, and are ideal for use in immunostaining of formalin fixed paraffin embedded (FFPE) and frozen tissue where stronger adhesion microscope slides are required. Glass Type: High Clarity, Low Flouorescence; Size: 75 x 25mm, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 45° Corner; Available Colors: White, Blue, Green, Yellow and Pink; Unit: 100 slides/box, 1,000 slides/case.
This item is not returnable. View return policy
Catalog No. NC2101954 Supplier Newcomer Supply Supplier No. 5080B
The surface of the TruBond 380 microscope slide is hydrophilic allowing the tissued to be refloated in the water bath for repositioning the tissue section and allowing the microscope slide to be used on all automated IHC platforms. They are especially useful in preventing tissue loss when performing high temperature antigen retrieval or enzyme digestion, and are ideal for use in immunostaining of formalin fixed paraffin embedded (FFPE) and frozen tissue where stronger adhesion microscope slides are required. Glass Type: High Clarity, Low Flouorescence; Size: 75 x 25mm, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 45° Corner; Available Colors: White, Blue, Green, Yellow and Pink; Unit: 100 slides/box, 1,000 slides/case.