Learn More
Electron Microscopy Sciences TruBond™ Fine Adhesion Slides, Green
SDP
Catalog No. 5034028
Encompass_Preferred
Adhesive slides have proven performance under high temperature and pH condition. These adhesive slides, including the hydrophilic TruBond 360, work well with automatic IHC applications. EMS is confident that the quality of these slides will exceed your expectations. For problems with tissue loss try our TruBond 380.
Color: Green
Catalog No. 50-340-28 Supplier Electron Microscopy Sciences Supplier No. 63703G1
Adhesive slides have proven performance under high temperature and pH condition. These adhesive slides, including the hydrophilic TruBond 360, work well with automatic IHC applications. EMS is confident that the quality of these slides will exceed your expectations. For problems with tissue loss try our TruBond 380.
Color: Green