Learn More
Covaris Inc TRUCHLP CHROMATIN W/ FORMA
SDP
Supplier:  Covaris Inc 520154
Encompass
The truChIP Chromatin Shearing Kit with Formaldehyde is designed and optimized for the efficient and reproducible shearing of chromatin from cultured mammalian cells (suspension or adherent) using the Covaris Adaptive Focused Acoustics (AFA) technology. Kit includes: Formaldehyde 16% Methanol-free Formaldehyde (5x1 mL ampules) Buffer A 7.5 mL 10X Fixing Buffer Buffer B 5 mL 5X Lysis Buffer Buffer C 2.5 mL 10X Wash Buffer Buffer D3 6 mL 10X SDS Shearing Buffer Buffer E 6 mL 1X Quenching Buffer Buffer F 0.8 mL 100X Protease Inhibitor Cocktail
Catalog No. NC0734532