Learn More
Perfex TruCLEAN Deluxe Disinfection
Catalog No. 19003139 Shop All Perfex Corp. Products
Change view
Click to view available options
Color:
Green
Orange
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-003-139 Orange
19-003-134 Red
19-003-135 White
19-003-136 -
19-003-137 Green
19-003-138 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-003-139 Supplier Perfex Supplier No. 303O
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quickly apply disinfecting agents onto floors, walls, and ceilings. Compact design for simplified use in confined areas.
Designed for quick application of disinfecting agents onto floors, walls, and ceilings. Simply dip the mop in solution, press mop on dosage sieve and apply to surface. Compact design is perfect for use in confined areas, allows easy storage with limited space.
Temperature-resistant 36 Liter polypropylene bucket includes embossed graduations for accurate measure and 3-inch twin-wheel casters with one locking caster. Easily accessible brake pedal allows parking on uneven floors. Stainless steel dosage sieve mounts evenly while precisely positioned apertures release excess disinfectant into 36 Liter or waste containment bucket.
Specifications
Color Orange
Dimensions 19 x 19 x 17 in.