Learn More
Perfex TruCLEAN Pro XL
Catalog No. 19005388 Shop All Perfex Corp. Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-005-388 Yellow
19-005-384 Red
19-005-385 White
19-005-386 Blue
19-005-387 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-005-388 Supplier Perfex Supplier No. 223XLY
Add to Cart
Add to Cart
This item is not returnable. View return policy
Triple 22 Liter bucket configuration with handle-mounted carrying basket, designed for large area cleaning and disinfecting.
Capture and isolate contaminants to ensure the delivery of unadulterated cleaning and sanitizing agents. No more re-applying dirty water and weakened solutions. The Pro XL system is designed to tackle very large area cleaning of floors, walls, and ceilings.
Equipped with large 22 Liter buckets and two 6 Liter pails in a handle-mounted carrying basket, the XL system easily manages your cleaning supplies with room to spare. Other features include a fold-away cart extension for easy storage and heavy-duty 4-inch casters for enhanced mobility.
Each mopping system component plays an important role in delivering the highest level of cleanliness. For example, our innovative swivel mop frame is accurately balanced to maintain total surface contact at any angle. The unique wringing mechanism exerts force evenly across the entire mop surface, extracting the maximum amount of soiled contaminants.
Specifications
Color Yellow
Dimensions (L x W x H) 40 x 19 x 23 in.