Learn More
StatLab™ TruCut™ Microtome Blades
GSA_VASDP
Catalog No. STLCUT7500
Change view
Click to view available options
Profile:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile
STLCUT7500 Low
STLCUT7550 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. STLCUT7500 Supplier StatLab™ Supplier No. CUT7500
Add to Cart
Add to Cart
This item is not returnable. View return policy
TruCut™ microtome blades demonstrate increased sharpness and greater durability as compared to many conventional microtome blades.
  • Blade edge is slowly refined, sharpened and optimized to maximize hardness, sharpness and durability
  • Made in Japan with a proprietary surface ionization process
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Profile Low
Disposable Single-use
Quantity 50 Pk.