Learn More
Stanbio™ True 20 Plus hCG Pregnancy Test Set
SDP
Catalog No. 22022632 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Type:
hCG Test
Catalog No. Type
22-022-632 hCG Test
1 options
This item is not returnable. View return policy
Catalog No. 22-022-632 Supplier Stanbio™ Supplier No. 1440050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Results in either three minutes or five minutes
  • Results in either three minutes (urine) or five minutes (serum)
  • 20 mIU/mL hCG or greater in urine or serum
Specifications
Type hCG Test