Learn More
Biotium TrueBlack IF Background Suppressor System, 200 assays, non-mammalian blocking agents, simultaneous blocking, permeabilization, antibody dilution, 10 min blocking time
Catalog No. NC1722154
Encompass_Preferred
TrueBlack IF Background Suppressor System, 200 assays, non-mammalian blocking agents, simultaneous blocking, permeabilization, antibody dilution, 10 min blocking time
Catalog No. NC1722154 Supplier Biotium Supplier No. 23012
TrueBlack IF Background Suppressor System, 200 assays, non-mammalian blocking agents, simultaneous blocking, permeabilization, antibody dilution, 10 min blocking time