Learn More
Electron Microscopy Sciences True Blue™ True Blue Film 8" Plain Back
SDP
Supplier:  Electron Microscopy Sciences 50336PB
Encompass_Preferred
Mixture of abrasive particle sizes, shapes bonded to film backing that enable film to produce finer surfaces than found in similar films. Film is typically used after 1 u diamond lapping film step after 1 micron diamond.
Backing: Plain
Diameter: 8"
Catalog No. 50-997-229