Learn More
Electron Microscopy Sciences True Blue™ True Blue Film 8" Plain Back
SDP
Catalog No. 501931671
Encompass_Preferred
True Blue™ film is produced with a proprietary process combining a unique mixture of abrasive particle sizes and shapes that are bonded to a film backing which enable the film to produce finer surfaces than are commonly found in similarly graded micron films. True Blue™ film is typically used after the 1 micron diamond lapping.
Backing: Plain
Diameter: 8"
Catalog No. 50-193-1671 Supplier Electron Microscopy Sciences Supplier No. 50336PB25
True Blue™ film is produced with a proprietary process combining a unique mixture of abrasive particle sizes and shapes that are bonded to a film backing which enable the film to produce finer surfaces than are commonly found in similarly graded micron films. True Blue™ film is typically used after the 1 micron diamond lapping.
Backing: Plain
Diameter: 8"