Learn More
Electron Microscopy Sciences True North™ 25 Microscope Slide Box, Blue
SDP
Catalog No. 501929499
Encompass_Preferred
Protect your slides in ultra-low temperature ranges down to -80 deg C with these stackable slide boxes. Safely store and transport 25 x 75 mm (3 x 1 inch) slides. The uniform grooved slide slots separate slides, preventing slide contact. They feature a temperature-resistant numbered foam lining and an identification sheet. Durable blue polycarbonate with a friction-fit lid.
Color: Blue
Capacity: 25 slides
Catalog No. 50-192-9499 Supplier Electron Microscopy Sciences Supplier No. 7148502
Protect your slides in ultra-low temperature ranges down to -80 deg C with these stackable slide boxes. Safely store and transport 25 x 75 mm (3 x 1 inch) slides. The uniform grooved slide slots separate slides, preventing slide contact. They feature a temperature-resistant numbered foam lining and an identification sheet. Durable blue polycarbonate with a friction-fit lid.
Color: Blue
Capacity: 25 slides