Learn More
Research Products International Corp True North Cool Container, Red, 9L
SDP
Catalog No. 501977626
Encompass
True North Cool Containers are made of seamless polyurethane for maximum durability and excellent insulating properties. Ideal for use with wet ice, dry ice or liquid nitrogen. Containers include a tight fitting lid that provides temperature stability and reduces evaporation. Unique design allows lid to conveniently store under the container.
Catalog No. 50-197-7626 Supplier Research Products International Corp Supplier No. 246403
True North Cool Containers are made of seamless polyurethane for maximum durability and excellent insulating properties. Ideal for use with wet ice, dry ice or liquid nitrogen. Containers include a tight fitting lid that provides temperature stability and reduces evaporation. Unique design allows lid to conveniently store under the container.