Learn More
Electron Microscopy Sciences True North™ 100 Microscope Slide Box, Purple
SDP
Catalog No. 50365742
Encompass_Preferred
Protect your slides in ultra-low temperature ranges down to -80 deg C with these stackable slide boxes. Safely store and transport 25 x 75 mm (3 x 1 inch) slides. The uniform grooved slide slots separate slides, preventing slide contact. They feature a temperature-resistant numbered foam lining and an identification sheet. Durable purple polycarbonate with a stainless steel clasp and nickel hinges.
Color: Purple
Capacity: 100 slides
Catalog No. 50-365-742 Supplier Electron Microscopy Sciences Supplier No. 7148501
Protect your slides in ultra-low temperature ranges down to -80 deg C with these stackable slide boxes. Safely store and transport 25 x 75 mm (3 x 1 inch) slides. The uniform grooved slide slots separate slides, preventing slide contact. They feature a temperature-resistant numbered foam lining and an identification sheet. Durable purple polycarbonate with a stainless steel clasp and nickel hinges.
Color: Purple
Capacity: 100 slides