Learn More
Analytical Sales & Services Analytical Sales TrueTaper 96-Well Collection Plate, 1 mL Volume, Maximum Sample Recovery
Catalog No. NC0749030
Encompass
The TrueTaper 96-Well Collection Plate offers superior sample recovery and minimizes dead volume with its unique tapered bottom design. Constructed from durable polypropylene and featuring SiliGuard™ Low Bind Coating, it ensures minimal sample loss and improved pipetting performance. Packaged in clamshells to prevent contamination, this product is ideal for precise analytical applications demanding high integrity and efficiency in sample handling.
Catalog No. NC0749030 Supplier Analytical Sales & Services Supplier No. 968810GC
The TrueTaper 96-Well Collection Plate offers superior sample recovery and minimizes dead volume with its unique tapered bottom design. Constructed from durable polypropylene and featuring SiliGuard™ Low Bind Coating, it ensures minimal sample loss and improved pipetting performance. Packaged in clamshells to prevent contamination, this product is ideal for precise analytical applications demanding high integrity and efficiency in sample handling.