Learn More
Biotium TRUEBLACK LIPO QUENCHER 30X
Catalog No. NC3622044
Encompass_Preferred
  • Effectively eliminates lipofuscin autofluorescence.
  • Reduces autofluorescence from other tissue components.
  • Can be incorporated before or after immunofluorescence staining.
  • Less toxic solvent compared to earlier formulations.
  • 30X concentration in DMSO for a more concentrated solution.
  • Improves fluorescence imaging in human and aged animal tissues.
  • Minimizes background fluorescence from red/far-red sources.
  • One mL treats approximately 150-300 tissue sections.
  • Stable for at least 12 months from receipt.
  • Store at room temperature, protect from light.
  • Heat vial to 70°C before dilution to avoid precipitates.
  • Optimized for imaging cytometry applications.
Catalog No. NC3622044 Supplier Biotium Supplier No. 23011
  • Effectively eliminates lipofuscin autofluorescence.
  • Reduces autofluorescence from other tissue components.
  • Can be incorporated before or after immunofluorescence staining.
  • Less toxic solvent compared to earlier formulations.
  • 30X concentration in DMSO for a more concentrated solution.
  • Improves fluorescence imaging in human and aged animal tissues.
  • Minimizes background fluorescence from red/far-red sources.
  • One mL treats approximately 150-300 tissue sections.
  • Stable for at least 12 months from receipt.
  • Store at room temperature, protect from light.
  • Heat vial to 70°C before dilution to avoid precipitates.
  • Optimized for imaging cytometry applications.