Learn More
Biotium TrueBlack WB Blocking Buffer Kit
Catalog No. 501964407
Encompass_Preferred
The TrueBlack WB blocking buffer is a buffer system designed to achieve optimal specificity and sensitivity by blocking background from the non-specific interaction between the fluorophore-labeled antibodies and the membrane with cell lysates. For blocking and antibody dilution for 50 membranes.
Catalog No. 50-196-4407 Supplier Biotium Supplier No. 23013
The TrueBlack WB blocking buffer is a buffer system designed to achieve optimal specificity and sensitivity by blocking background from the non-specific interaction between the fluorophore-labeled antibodies and the membrane with cell lysates. For blocking and antibody dilution for 50 membranes.