Learn More
Rockland Immunochemicals TrueBlot Anti-Rabbit Ig IP Agarose Beads - 00-8800-25
SDP
Catalog No. NC0410568
Encompass_Preferred
TrueBlot Anti-Rabbit Ig IP Agarose Beads are a suspension of activated agarose beads coupled with goat Anti-rabbit IgG. It is suitable for precipitation of rabbit IgGs used as the primary antibodies in immunoprecipitation assays. The beads are in suspension and will settle upon storage. Prior to use, mix the vial gently (do not vortex) to ensure delivery of proper bead volume.
Catalog No. NC0410568 Supplier Rockland Immunochemicals Supplier No. 00880025
TrueBlot Anti-Rabbit Ig IP Agarose Beads are a suspension of activated agarose beads coupled with goat Anti-rabbit IgG. It is suitable for precipitation of rabbit IgGs used as the primary antibodies in immunoprecipitation assays. The beads are in suspension and will settle upon storage. Prior to use, mix the vial gently (do not vortex) to ensure delivery of proper bead volume.